Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methoxyanthranilic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Amino-3-methoxybenzoic acid3-Methoxyanthranilic acido-Anisidine-6-carboxylic acid2-Amino-m-anisic acid

43772-0250 43772-0050 43772-0010
TS43772-0250EA 559 USD InStock
TS43772-0250 TS43772-0050 TS43772-0010
3-Methoxyanthranilic acid 98%
3-Methoxyanthranilic acid


Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 168…170.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00075178
CAS Number: 3177-80-8
Order Now
Specification Test Results
Appearance White to yellow to brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty