Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xylene Cyanole FF ∼75% ~75%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium hydrogen 4-[[4-(ethylamino)-m-tolyl][4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl]benzene-1,3-disulfonateSodium 4-((4-(ethylamino)-3-methylphenyl)(4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-sulfobenzenesulfonateacid Blue 147Sodium 4-((4-(ethylamino)-3-methylphenyl)(4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-sulphobenzenesulphonateSodium hydrogen 4-[[4-(ethylamino)-m-tolyl][4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl]benzene-1,3-disulphonateC.I. 42135

44070-0100 44070-0500
TS44070-0500EA 198 USD InStock
TS44070-0500 TS44070-0100
Xylene Cyanole FF ∼75% ~75%
Xylene Cyanole FF


Formula: C₂₅H₂₇N₂NaO₆S₂
MW: 538.62 g/mol
Melting Pt: 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00040657
CAS Number: 2650-17-1
Order Now
Specification Test Results
Appearance Blue-grey to green Powder
Infrared spectrum Conforms
Loss on drying ≤5 % (105°C)
Solubility (0.1% in water) Dark blue
Dye content ≥70 %
UV Lambda max 612 to 616 nm
Suitability (as track. dye for DNA seq) Passes test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty