Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium sulfate decahydrate 99% for biochemistry

Supplier: Thermo Scientific Chemicals

44793-5000 44793-0025
TS44793-0025EA 252 USD InStock
TS44793-0025 TS44793-5000
Sodium sulfate decahydrate 99% for biochemistry
Sodium sulphate decahydrate


Formula: Na₂SO₄·10H₂O
MW: 322.2 g/mol
Melting Pt: 884 °C
Storage Temperature: Ambient
MDL Number: MFCD00149189
CAS Number: 7727-73-3
Order Now
Specification Test Results
Appearance White Crystalline powder
Loss on drying 52 % to 57 %
Solubility Clear colorless
UV (1 M in water)
UV at 260 nm A: ≤0.025
UV at 280 nm A: ≤0.021
pH 5.2 to 9.2 (5 % in water)
Titration after Ion exchange ≥98.5 %
Chloride (Cl) ≤0.02 %
Arsenic (As) ≤0.0001 %
Calcium (Ca) ≤0.02 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.01 %
Heavy metals (as Pb) ≤0.001 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty