Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanolamine ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Monoethanolamine2-Aminoethanol

45176-0010 45176-2500 45176-0025
TS45176-0010EA 82.17 USD InStock
TS45176-0010 TS45176-0025 TS45176-2500
Ethanolamine ACS
Ethanolamine


Formula: NH₂CH₂CH₂OH
MW: 61.08 g/mol
Boiling Pt: 170 °C (760 mmHg)
Melting Pt: 10.3 °C
Density: 1.012 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00008183
CAS Number: 141-43-5
UN: 2491
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.30 %
GC ≥99.0 %
Color scale ≤15 APHA
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty