Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fusidic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

45846-0010 45846-0050
TS45846-0010EA 100 USD InStock
TS45846-0010 TS45846-0050
Fusidic acid 98%
Fusidic acid


Formula: C₃₁H₄₈O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00865135
CAS Number: 6990-06-3
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤10 %
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty