Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Irinotecan hydrochloride trihydrate

Supplier: Thermo Scientific Chemicals
Warning

46154-2500 46154-0010
TS46154-0010EA 728 USD InStock
TS46154-0010 TS46154-2500
Irinotecan hydrochloride trihydrate
Irinotecan hydrochloride trihydrate


Formula: C₃₃H₃₈N₄O₆·HCl·3H₂O
MW: 677.19 g/mol (623.15 anhy)
Storage Temperature: Freezer
MDL Number: MFCD01765731
CAS Number: 136572-09-3
Order Now
Specification Test Results
Appearance Yellow Powder
Infrared spectrum Conforms
Water 7 to 9 %
HPLC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty