Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Aminobutyric acid 99+%

Supplier: Thermo Scientific Chemicals
Synonyms: γ-Aminobutyric acidGABA

10328-5000 10328-1000 10328-0250 10328-0025
TS10328-0025EA 807 USD InStock
TS10328-0025 TS10328-5000 TS10328-1000 TS10328-0250
4-Aminobutyric acid 99+%
4-Aminobutyric acid


Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: 197…204 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008226
CAS Number: 56-12-2
EINECS: 200-258-6
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 197 to 204 °C
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %
UV transmittance ≥95 % (c=10, 2 N HCl, 430 nm) 1 cm cell
Titration with HClO4 ≥99.0 % (On anhydrous substance)
Chloride (Cl) ≤400 ppm
Sulfate (SO4) ≤500 ppm
Heavy metals (as Pb) ≤10 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty