Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AMP (2-Amino-2-methylpropanol) 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Amino-2-methyl-1-propanolAMP

10406-0050 10406-0010 10406-5000
TS10406-0010EA 66.1 USD InStock
TS10406-0010 TS10406-5000 TS10406-0050
AMP (2-Amino-2-methylpropanol) 99%
AMP (2-Amino-2-methylpropanol)


Formula: (CH₃)₂C(NH₂)CH₂OH
MW: 89.14 g/mol
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: 31…32 °C
Density: 0.93 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00008051
CAS Number: 124-68-5
EINECS: 204-709-8
Order Now
Specification Test Results
Appearance Colorless Low melting solid
Infrared spectrum Conforms
Water ≤0.8 %
Titration with HClO4 ≥98.5 %
UV 10 cm cell vs water
UV at 420 nm A: ≤0.005
Color scale ≤10 APHA
Suitability (in liquid scint. counting) Passes test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty