Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-2-naphthoic acid 85% tech.

Supplier: Thermo Scientific Chemicals
Warning

10414-0250 10414-0050
TS10414-0050EA 131 USD InStock
TS10414-0050 TS10414-0250
3-Amino-2-naphthoic acid 85% tech.
3-Amino-2-naphthoic acid


Formula: C₁₁H₉NO₂
Boiling Pt: 383 °C (1013 hPa)
Melting Pt: 210 °C (dec.)
MDL Number: MFCD00004115
CAS Number: 5959-52-4
EINECS: 227-726-2
Order Now
Specification Test Results
Appearance Yellow to green Powder
Infrared spectrum Conforms
HPLC ≥80 %
Titration with HClO4 ≥80 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty