Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-1-phenylethanol 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: PhenylethanolamineDL-β-Hydroxyphenethylaminealpha-(Aminomethyl)benzyl Alcohol

10432-0500 10432-0100
TS10432-0100EA 107 USD InStock
TS10432-0100 TS10432-0500
2-Amino-1-phenylethanol 98%
2-Amino-1-phenylethanol
Appearance: Clear yellow Viscous liquid


Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 160 °C (17 mmHg)
Melting Pt: 56…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008137
CAS Number: 7568-93-6
UN: 3259
ADR: 8,III
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 52 to 60 °C
HPLC ≥97.5 %
Specific optical rotation +200° to +220° (20 °C, 589 nm) (c=3, CHCl3)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty