Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butylamine 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Aminoisobutane

10782-0025 10782-0010 10782-0050 10782-0100
TS10782-0050EA 27 USD InStock
TS10782-0050 TS10782-0025 TS10782-0010 TS10782-0100
tert-Butylamine 99%
tert-Butylamine


Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 46 °C (760 mmHg)
Melting Pt: –67 °C
Density: 0.69 g/cm³
Flash Pt: –38 °C
Storage Temperature: Ambient
MDL Number: MFCD00008050
CAS Number: 75-64-9
EINECS: 200-888-1
UN: 2734
ADR: 8,I
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3770 to 1.3790 (20°C, 589 nm)
Water ≤0.3 %
GC ≥98.5 %
Color scale ≤10 APHA
SO4 and SO3 (as SO4) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Titrable acid ≤0.002 meq/g
Insoluble matter ≤0.005 %
Starch Passes test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty