Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Butyronitrile 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: n-Butyronitrile

10814-0025 10814-1000 10814-5000
TS10814-0025EA 519 USD InStock
TS10814-0025 TS10814-1000 TS10814-5000
Butyronitrile 99%
Butyronitrile


Formula: C₄H₇N
MW: 69.11 g/mol
Boiling Pt: 115…117 °C (760 mmHg)
Melting Pt: –112 °C
Density: 0.79 g/cm³
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00001968
CAS Number: 109-74-0
EINECS: 203-700-6
UN: 2411
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3830 to 1.3850 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤25 APHA
UV at 250 nm A: ≤0.005
Color scale ≤5 APHA
Residue after evaporation ≤2 ppm
Acidity ≤0.0002 meq/g

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty