Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Diaminopropane 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,3-PropanediamineTrimethylenediamine

11235-0010 11235-0500 11235-2500
TS11235-0010EA 148.31 USD InStock
TS11235-0010 TS11235-2500 TS11235-0500
1,3-Diaminopropane 99%
1,3-Diaminopropane


Formula: C₃H₁₀N₂
MW: 74.13 g/mol
Boiling Pt: 140 °C (760 mmHg)
Melting Pt: –12 °C
Density: 0.88 g/cm³
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00008228
CAS Number: 109-76-2
EINECS: 203-702-7
UN: 2734
ADR: 8,I
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4565 to 1.4585 (20°C, 589 nm)
Water ≤0.5 %
GC ≥98.5 %
Color scale ≤20 APHA
Titration with HClO4 ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty