Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Diethyl-m-toluamide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: DiethyltoluamideDeetDiethyl-m-toluamideN,N-Diethyl-3-methylbenzamide

11457-0025 11457-1000 11457-5000
TS11457-0025EA 211 USD InStock
TS11457-0025 TS11457-5000 TS11457-1000
N,N-Diethyl-m-toluamide 98%
N,N-Diethyl-m-toluamide


Formula: C₁₂H₁₇NO
MW: 191.27 g/mol
Boiling Pt: 111 °C (1 mmHg)
Density: 0.9955 g/cm³
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00009046
CAS Number: 134-62-3
EINECS: 205-149-7
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5200 to 1.5240 (25°C, 589 nm)
Water ≤0.3 %
GC ≥97.5 %
Color scale ≤100 APHA

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty