Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diaminomaleonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

17270-5000 17270-0250 17270-1000
TS17270-0250EA 35 USD InStock
TS17270-0250 TS17270-1000 TS17270-5000
Diaminomaleonitrile 98%
Diaminomaleonitrile


Formula: C₄H₄N₄
MW: 108.1 g/mol
Boiling Pt: 445 °C (1013 hPa)
Melting Pt: 178…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00001870
CAS Number: 1187-42-4
EINECS: 214-697-6
UN: 3439
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Brown Crystalline powder
Infrared spectrum Conforms
UV spectrophotometry ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty