Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Phenylalaninol 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: L(-)-2-Amino-3-phenylpropan-1-ol(S)-(-)-Phenylalaninol(S)-2-Amino-3-phenylpropan-1-olL(-)-2-Amino-3-phenyl-1-propanolH-Phenylalaninol(S)-(-)-2-Amino-3-phenyl-1-propanol(S)-(-)-2-Amino-3-phenylpropan-1-olL(-)-Phenylalaninol

18387-0250 18387-0050 18387-0010
TS18387-0250EA 481 USD InStock
TS18387-0250 TS18387-0050 TS18387-0010
L-Phenylalaninol 98%
L-Phenylalaninol


Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00004732
CAS Number: 3182-95-4
EINECS: 221-674-4
UN: 3259
ADR: 8,III
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 90 to 94 °C
Specific optical rotation -21° to -25° (22 °C, 589 nm) (c=1.2, 1 N HCl)
Titration with HClO4 ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty