Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclobutylamine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: CyclobutanamineAminocyclobutaneCyclobutylamine

20911-0050 20911-0010
TS20911-0050EA 182 USD InStock
TS20911-0050 TS20911-0010
Cyclobutylamine 98%
Cyclobutylamine


Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 81.4 °C (1013 hPa)
Density: 0.83 g/cm³
Flash Pt: –4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001328
CAS Number: 2516-34-9
EINECS: 219-736-0
UN: 2733
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4360 to 1.4400 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty