Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,8-Diaminonaphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Naphthalene-1,8-diamine1,8-Naphthalenediamine1,8-naphthylenediamine

37118-0050 37118-5000 37118-1000
TS37118-5000EA 319 USD InStock
TS37118-5000 TS37118-0050 TS37118-1000
1,8-Diaminonaphthalene 97%
1,8-Diaminonaphthalene


Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 205 °C (12 mmHg)
Melting Pt: 63…66 °C
Flash Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00004033
CAS Number: 479-27-6
EINECS: 207-529-8
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Grey to red to purple to dark brown Crystals or flakes and/or chunks
Infrared spectrum Conforms
Melting point 61 to 66 °C
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty