Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-1,3-propanediol 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Serinol2-Amino-1,3-dihydroxy propane2-Aminopropane-1,3-diol

40103-0250 40103-0050
TS40103-0050EA 59 USD InStock
TS40103-0050 TS40103-0250
2-Amino-1,3-propanediol 98%
2-Amino-1,3-propanediol


Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 115…116 °C (760 mmHg)
Melting Pt: 52…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051487
CAS Number: 534-03-2
EINECS: 208-584-0
UN: 3259
ADR: 8,III
Order Now
Specification Test Results
Appearance White to off-white Adhering crystalline powder
Infrared spectrum Conforms
Melting point 52 to 56 °C
Titration with HClO4 ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty