Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(-)-2-Amino-3-methyl-1-butanol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (R)-(-)-2-Amino-3-methylbutanolD(-)-2-Amino-3-methyl-1-butanolD(-)-ValinolD-Valinol

42247-0050 42247-0250
TS42247-0250EA 714 USD InStock
TS42247-0250 TS42247-0050
(R)-(-)-2-Amino-3-methyl-1-butanol 98%
(R)-(-)-2-Amino-3-methyl-1-butanol


Formula: C₅H₁₃NO
MW: 103.16 g/mol
Melting Pt: 32…36 °C
Density: 0.931 g/cm³
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064297
CAS Number: 4276-09-9
Order Now
Specification Test Results
Appearance White to light yellow Crystals or low melting crystalline mass
Infrared spectrum Conforms
Melting point 32 to 36 °C
GC ≥97.5 %
Specific optical rotation -15° to -18° (20 °C, 589 nm) (c=10, EtOH)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty