Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allantoin 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-(2,5-Dioxoimidazolidin-4-yl)urea

10284-5000 10284-1000
TS10284-5000EA 140.99 USD InStock
TS10284-5000 TS10284-1000
Allantoin 98%
Allantoin


Formula: C₄H₆N₄O₃
MW: 158.12
Melting Pt: 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005260
CAS Number: 97-59-6
EINECS: 202-592-8
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
Titration with HClO4 ≥98.5 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty