Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-3-amino-1H-pyrazole-4-carboxylate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 3-Amino-4-carbethoxypyrazoleEthyl 3-amino-1H-pyrazole-4-carboxylate

10331-0050
TS10331-0050EA 65 USD InStock
TS10331-0050
Ethyl-3-amino-1H-pyrazole-4-carboxylate 99%
Ethyl 3-amino-1H-pyrazole-4-carboxylate


Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00005238
CAS Number: 6994-25-8
EINECS: 230-262-3
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
HPLC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty