Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorophenethylamine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(4-Chlorophenyl)ethylamine

10988-0050 10988-0250
TS10988-0250EA 85 USD InStock
TS10988-0250 TS10988-0050
4-Chlorophenethylamine 98%
4-Chlorophenethylamine


Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Boiling Pt: 242…245 °C (760 mmHg)
Density: 1.112 g/cm³
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00008191
CAS Number: 156-41-2
EINECS: 205-853-4
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5474 to 1.5494 (20 °C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty