Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Ornithine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Orn-OH·HClH-Orn-OH Hydrochloride

12956-1000 12956-5000 12956-0250
TS12956-0250EA 46 USD InStock
TS12956-0250 TS12956-1000 TS12956-5000
L(+)-Ornithine hydrochloride 99%
L(+)-Ornithine hydrochloride


Formula: C₅H₁₂N₂O₂·HCl
MW: 168.62 g/mol
Melting Pt: 245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064562
CAS Number: 3184-13-2
EINECS: 221-678-6
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Solubility (5 % in 1 N HCl) Clear colorless
Specific optical rotation +21° to +24° (20 °C, 589 nm) (c=5, 1 N HCl)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty