Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitrilotriacetic acid trisodium salt monohydrate 99+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Nitrilotriacetic acid, trisodium salt, monohydrate

14784-0250 14784-0025 14784-5000
TS14784-0025EA 193 USD InStock
TS14784-0025 TS14784-0250 TS14784-5000
Nitrilotriacetic acid trisodium salt monohydrate 99+%
Nitrilotriacetic acid trisodium salt monohydrate


Formula: C₆H₆NNa₃O₆·1H₂O
MW: 275.1 g/mol
Melting Pt: >320 °C
Storage Temperature: Ambient
MDL Number: MFCD00150789
CAS Number: 18662-53-8
EINECS: 225-768-6
Order Now
Specification Test Results
Appearance White Powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with HClO4 ≥99.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty