Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Betaine hydrochloride 99%, Extra pure meets analytical specification of USP

Supplier: Thermo Scientific Chemicals
Warning

16455-0025 16455-0010 16455-2500
TS16455-0025EA 227 USD InStock
TS16455-0025 TS16455-2500 TS16455-0010
Betaine hydrochloride 99%, Extra pure meets analytical specification of USP
Betaine hydrochloride


Formula: C₅H₁₂ClNO₂
MW: 153.61
Melting Pt: 241 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011903
CAS Number: 590-46-5
Order Now
Specification Test Results
Appearance Colorless to white Powder or crystals
Infrared spectrum Conforms (identification A)
Water ≤0.5 %
Identification B Passes test
Residue after ignition ≤0.1 %
pH 0.8 to 1.2 (1 in 4 solution)
Titration with HClO4 98.0 to 100.5 % (On anhydrous basis)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty