Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetamidine hydrochloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

17021-5000 17021-1000 17021-0100
TS17021-1000EA 60 USD InStock
TS17021-1000 TS17021-0100 TS17021-5000
Acetamidine hydrochloride 98+%
Acetamidine hydrochloride


Formula: C₂H₇ClN₂
Melting Pt: 160…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00013016
CAS Number: 124-42-5
EINECS: 204-700-9
Order Now
Specification Test Results
Appearance White to cream Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric 98.0 to 102.0 %
HPLC 95.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty