Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-7-nitrobenzofurazan 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: NBD-ClNBD chloride4-Chloro-7-Nitrobenz-2-Oxa-1,3-Diazole

17239-0010 17239-0250 17239-0050
TS17239-0050EA 71 USD InStock
TS17239-0050 TS17239-0250 TS17239-0010
4-Chloro-7-nitrobenzofurazan 98%
4-Chloro-7-nitrobenzofurazan


Formula: C₆H₂ClN₃O₃
MW: 199.55 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00005808
CAS Number: 10199-89-0
EINECS: 233-496-4
Order Now
Specification Test Results
Appearance Yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 96 to 99 °C
Sulfated ash ≤0.1 %
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty