Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(1-Naphthylmethyl)-2-imidazoline hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: AlbalonNaphazoline Hydrochloride

17966-1000 17966-0250
TS17966-1000EA 168 USD InStock
TS17966-1000 TS17966-0250
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride 99%
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride


Formula: C₁₄H₁₅ClN₂
MW: 246.738 g/mol
Melting Pt: 254…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00012554
CAS Number: 550-99-2
EINECS: 208-989-2
UN: 2811
ADR: 6.1,II
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty