Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-2-piperidinecarboxylate 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ethyl pipecolinate

18958-0250 18958-0050
TS18958-0250EA 165 USD InStock
TS18958-0250 TS18958-0050
Ethyl-2-piperidinecarboxylate 98+%
Ethyl 2-piperidinecarboxylate


Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Density: 1.006 g/cm³
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005980
CAS Number: 15862-72-3
EINECS: 239-990-6
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4545 to 1.4575 (20 °C, 589 nm)
GC ≥98.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty