Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butyl-N-hydroxycarbamate 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: N-tert-ButoxycarbonylhydroxylamineN-Tert-Boc-HydroxylamineN-Boc-hydroxylamine

20987-0010 20987-0050 20987-0250
TS20987-0050EA 95 USD InStock
TS20987-0050 TS20987-0250 TS20987-0010
tert-Butyl-N-hydroxycarbamate 98+%
tert-Butyl N-hydroxycarbamate


Formula: C₅H₁₁NO₃
MW: 133.15 g/mol
Melting Pt: 53…57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002107
CAS Number: 36016-38-3
EINECS: 252-836-2
Order Now
Specification Test Results
Appearance White to light pink Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 57 °C
GC ≥98.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty