Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D(+)-Ribonic acid γ-lactone 96%

Supplier: Thermo Scientific Chemicals
Synonyms: D-Ribonic acid-1,4-lactoneRibono-γ-lactone(3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one

22783-0100
TS22783-0100EA 294 USD InStock
TS22783-0100
D(+)-Ribonic acid γ-lactone 96%
D(+)-Ribonic acid γ-lactone


Formula: C₅H₈O₅
MW: 148.11 g/mol
Melting Pt: 83…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063241
CAS Number: 5336-08-3
EINECS: 226-256-5
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
GC ≥95 %
Residue after ignition ≤0.2 %
Specific optical rotation +17° to +19° (20 °C, 589 nm) (c=5, H2O)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty