Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methoxyphenethylamine 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(3-Methoxyphenyl)ethylamine

30261-0050 30261-0250
TS30261-0050EA 114 USD InStock
TS30261-0050 TS30261-0250
3-Methoxyphenethylamine 98+%
3-Methoxyphenethylamine


Formula: C₉H₁₃NO
Boiling Pt: 118…119 °C (6 mmHg)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008187
CAS Number: 2039-67-0
EINECS: 218-017-9
UN: 2735
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5375 to 1.5395 (20 °C, 589 nm)
GC ≥98.0 %
Titration with HClO4 ≥98.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty