Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-4-amino-3-chlorobenzoate 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methyl 4-amino-3-chlorobenzoate

32639-0010 32639-0050
TS32639-0050EA 157 USD InStock
TS32639-0050 TS32639-0010
Methyl-4-amino-3-chlorobenzoate 97%
Methyl 4-amino-3-chlorobenzoate


Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Melting Pt: 106…110 °C
Storage Temperature: Ambient
MDL Number: MFCD01320686
CAS Number: 84228-44-4
Order Now
Specification Test Results
Appearance Beige to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 106 to 110 °C
HPLC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty