Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(-)-4-Amino-2-hydroxybutyric acid 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (S)-4-Amino-2-hydroxybutanoic acid

34243-0050 34243-0250
TS34243-0250EA 197 USD InStock
TS34243-0250 TS34243-0050
(S)-(-)-4-Amino-2-hydroxybutyric acid 96%
(S)-(-)-4-Amino-2-hydroxybutyric Acid


Order Now
Specification Test Results
Appearance White to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Specific optical rotation -28° to -32° (20 °C, 589 nm) (c=1, water)
Titration with HClO4 ≥95 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty