Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-L-prolinol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (S)-1-Boc-2-pyrrolidinemethanol(S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanolN-Boc-L-ProlinolN-(tert-Butoxycarbonyl)-L-prolinol

34456-0050 34456-0250
TS34456-0250EA 505 USD InStock
TS34456-0250 TS34456-0050
N-Boc-L-prolinol 98%
N-Boc-L-prolinol


Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00066232
CAS Number: 69610-40-8
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 58 to 63 °C
GC ≥97.5 %
Specific optical rotation -46° to -52° (20 °C, 589 nm) (c=1, CHCl3)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty