Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluorophenethylamine 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(3-Fluorophenyl)ethylamine

35094-0050 35094-0250
TS35094-0250EA 283 USD InStock
TS35094-0250 TS35094-0050
3-Fluorophenethylamine 99%
3-Fluorophenethylamine


Formula: C₈H₁₀FN
Boiling Pt: 87 °C (15 mmHg)
Density: 1.066 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00075376
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5080 to 1.5100 (20 °C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty