Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-6-aminonicotinate 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methyl 6-aminonicotinate6-Aminonicotinic acid methyl ester

36681-0050 36681-0250
TS36681-0050EA 149 USD InStock
TS36681-0050 TS36681-0250
Methyl-6-aminonicotinate 97%
Methyl 6-aminonicotinate


Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 159…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00797844
CAS Number: 36052-24-1
Order Now
Specification Test Results
Appearance White to yellow to light beige to pink Powder
Infrared spectrum Conforms
Melting point 155 to 160 °C
HPLC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty