Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

N,N,N',N',N''-Pentamethyldiethylenetriamine 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,1,4,7,7-PentamethyldiethylenetriamineN1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine

41695-5000 41695-1000 41695-0250
TS41695-5000EA 182 USD InStock
TS41695-5000 TS41695-0250 TS41695-1000
N,N,N',N',N''-Pentamethyldiethylenetriamine 98+%
N,N,N',N',N''-Pentamethyldiethylenetriamine


Formula: C₉H₂₃N₃
MW: 173.30 g/mol
Melting Pt: 18 °C
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00014876
CAS Number: 3030-47-5
EINECS: 221-201-1
UN: 2922
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4415 to 1.4435 (20 °C, 589 nm)
GC ≥98.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty