Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Phenylalaninol 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: D-PenylalaninolD(+)-2-Amino-3-phenyl-1-propanol

41734-0010 41734-0100
TS41734-0100EA 295 USD InStock
TS41734-0100 TS41734-0010
D-Phenylalaninol 98%
D-Phenylalaninol


Formula: C₉H₁₃NO
MW: 151.2 g/mol
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00064298
CAS Number: 5267-64-1
EINECS: 226-086-1
UN: 3259
ADR: 8,III
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 90 to 94 °C
HPLC ≥97.5 %
Specific optical rotation +21° to +25° (22 °C, 589 nm) (c=1.2, 1 N HCl)
Titration with HClO4 ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty