Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-5-iodo-2-nitroaniline 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR55450-1G OR55450-250MG
76592-510EA 567 USD InStock
76592-510 76592-508
4-Bromo-5-iodo-2-nitroaniline 95%
4-Bromo-5-iodo-2-nitroaniline


Formula: C₆H₄BrIN₂O₂
MW: 342.917 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31543536
CAS Number: 2090498-07-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty