Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Valeric acid 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Pentanoic acid

OR55656-25G OR55656-100G
76621-696EA 13.39 USD InStock
76621-696 76621-694
Valeric acid 99%
Valeric acid


Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 184…186 °C
Melting Pt: –34 °C
Density: 0.938 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004413
CAS Number: 109-52-4
UN: 3265
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty