Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methyl-3-nitrobenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR5672-1G
76592-924EA 13.39 USD InStock
76592-924
4-Methyl-3-nitrobenzaldehyde 98%
4-Methyl-3-nitrobenzaldehyde


Formula: C₈H₇NO₃
Melting Pt: 44…49 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00017011
CAS Number: 31680-07-6
EINECS: 250-760-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty