Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyruvaldehyde (methylglyoxal) 0.4 in aqueous solution

Supplier: APOLLO SCIENTIFIC
Danger

OR5752-100G OR5752-500G OR5752-1KG
76593-020EA 101 USD InStock
76593-020 76593-022 76593-018
Pyruvaldehyde (methylglyoxal) 0.4 in aqueous solution
Pyruvaldehyde (Methylglyoxal)


Formula: C₃H₄O₂
Melting Pt: –20 °C
Density: 1.18 g/cm³ (25 °C)
Flash Pt: >71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006960
CAS Number: 78-98-8
EINECS: 201-164-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty