Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Dimethoxyphenylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Dihydroxy(2,5-dimethoxyphenyl)borane(2,5-Dimethoxyphenyl)metaboric acid2,5-Dimethoxybenzeneboronic acid(2,5-Dimethoxyphenyl)boronic acid

OR5823-100G OR5823-25G OR5823-5G
76593-082EA 79 USD InStock
76593-082 76593-084 76593-080
2,5-Dimethoxyphenylboronic acid
2,5-Dimethoxyphenylboronic acid


Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
Melting Pt: 95…98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318181
CAS Number: 107099-99-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty