Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butylchlorodiphenylsilane

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: tert-Butyldiphenylchlorosilanetert-Butyl-diphenylchlorosilane

OR5837-100G OR5837-500G OR5837-25G
76617-040EA 24.72 USD InStock
76617-040 76617-042 76617-038
tert-Butylchlorodiphenylsilane
tert-Butylchlorodiphenylsilane


Formula: C₁₆H₁₉ClSi
MW: 274.87 g/mol
Boiling Pt: 90…92 °C/0.01mmHg
Density: 1.057 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000497
CAS Number: 58479-61-1
EINECS: 261-282-0
UN: 2987
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty