Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminopyrimidine

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Pyrimidin-2-aminePyrimidin-2-ylamine

OR5847-100G OR5847-500G
76593-136EA 28 USD InStock
76593-136 76593-138
2-Aminopyrimidine
2-Aminopyrimidine


Formula: C₄H₅N₃
MW: 95.1 g/mol
Boiling Pt: 158 °C/186mmHg
Melting Pt: 122…126 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00006089
CAS Number: 109-12-6
EINECS: 203-648-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty