Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl isothiocyanatoformate 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Ethoxycarbonyl isothiocyanate

OR59819-100G OR59819-25G OR59819-5G
76612-168EA 31 USD InStock
76612-168 76612-166 76612-164
Ethyl isothiocyanatoformate 98%
Ethyl isothiocyanatoformate


Formula: C₄H₅NO₂S
MW: 131.16 g/mol
Boiling Pt: 56 °C/18mmHg
Density: 1.112 g/cm³ (25 °C)
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004814
CAS Number: 16182-04-0
EINECS: 240-318-9
UN: 1992
ADR: 3+6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty