Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclobutylamine

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: CyclobutanamineAminocyclobutaneCyclobutylamine

OR59991-25G OR59991-5G OR59991-1G
76612-188EA 76 USD InStock
76612-188 76612-186 76612-184
Cyclobutylamine
Cyclobutylamine


Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 81…83 °C
Density: 0.833 g/cm³ (25 °C)
Flash Pt: –4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001328
CAS Number: 2516-34-9
EINECS: 219-736-0
UN: 2733
ADR: 3+8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty