Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Iodobenzyl alcohol

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (4-Iodophenyl)methanol

OR60044-5G OR60044-10G OR60044-25G
76593-620EA 87 USD InStock
76593-620 76593-618 76593-616
4-Iodobenzyl alcohol
4-Iodobenzyl alcohol


Formula: C₇H₇IO
MW: 234.04 g/mol
Melting Pt: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD01732720
CAS Number: 18282-51-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty